Assay Details
| Gene Symbol: SOCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097461.1 | NP_001090930.1 | 2 | 546 | 71 | |
| XM_005664258.3 | XP_005664315.1 | 928 | 71 | |||
| XM_005664259.3 | XP_005664316.1 | 1220 | 71 | |||
| XM_005664260.3 | XP_005664317.1 | 1095 | 71 | |||
| XM_013998080.2 | XP_013853534.1 | 802 | 71 | |||
| XM_013998081.2 | XP_013853535.1 | 778 | 71 | |||
| XM_013998083.2 | XP_013853537.1 | 776 | 71 | |||
| XM_013998084.2 | XP_013853538.1 | 750 | 71 | |||
| GenBank mRNA | AK397161.1 | - | 3 | 890 | 71 | |
| EF121242.1 | - | 2 | 546 | 71 | ||
| HM045421.1 | - | 3 | 687 | 71 | ||
More Information
Set Membership: |
|

SOCS2
Genomic Map