Assay Details
| Gene Symbol: SLIRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097472.1 | NP_001090941.1 | 3 | 254 | 111 | |
| GenBank mRNA | AK400022.1 | - | 3 | 257 | 111 | |
| DQ629150.1 | - | 3 | 254 | 111 | ||
More Information
Set Membership: |
|

SLIRP
Genomic Map