Assay Details
| Gene Symbol: HPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098588.1 | NP_001092058.1 | 17 | 2947 | 123 | |
| XM_021068173.1 | XP_020923832.1 | 2859 | 123 | |||
| GenBank mRNA | AB271942.1 | - | 17 | 2947 | 123 | |
More Information
Set Membership: |
|

HPS3
Genomic Map