Assay Details
| Gene Symbol: IFIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100194.1 | NP_001093664.1 | 16 | 2955 | 126 | |
| XM_005671881.3 | XP_005671938.1 | 3146 | 126 | |||
| GenBank mRNA | EU006039.1 | - | 16 | 2955 | 126 | |
More Information
Set Membership: |
|

IFIH1
Genomic Map