Assay Details
| Gene Symbol: INS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109772.1 | NP_001103242.1 | 2 | 150 | 96 | |
| XM_021081278.1 | XP_020936937.1 | 361 | 96 | |||
| GenBank mRNA | AF064555.1 | - | 2 | 150 | 96 | |
More Information
Set Membership: |
|

INS
Genomic Map