Assay Details
| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112690.1 | NP_001106160.1 | 5 | 845 | 91 | |
| XM_021066031.1 | XP_020921690.1 | 788 | 91 | |||
| XM_021066032.1 | XP_020921691.1 | 1394 | 91 | |||
| GenBank mRNA | EF591082.1 | - | 5 | 845 | 91 | |
More Information
Set Membership: |
|

TNFSF13
Genomic Map