Assay Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112691.1 | NP_001106161.1 | 7 | 650 | 66 | |
| XM_021066033.1 | XP_020921692.1 | 611 | 66 | |||
| GenBank mRNA | EF624456.1 | - | 7 | 650 | 66 | |
More Information
Set Membership: |
|

TNFSF12
Genomic Map