Assay Details
| Gene Symbol: CIDEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112696.2 | NP_001106166.1 | 5 | 548 | 75 | |
| XM_005656107.3 | XP_005656164.1 | 936 | 75 | |||
| XM_005656108.3 | XP_005656165.1 | 608 | 75 | |||
| GenBank mRNA | EF394366.1 | - | 5 | 550 | 75 | |
More Information
Set Membership: |
|

CIDEA
Genomic Map