Assay Details
| Gene Symbol: CHORDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113446.1 | NP_001106917.1 | 10 - 11 | 845 | 123 | |
| GenBank mRNA | EU295560.1 | - | 10 - 11 | 845 | 123 | |
More Information
Set Membership: |
|

CHORDC1
Genomic Map