Assay Details
| Gene Symbol: GUSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123121.1 | NP_001116593.1 | 12 | 1853 | 62 | |
| GenBank mRNA | AK232674.1 | - | 12 | 1855 | 62 | |
| AK392337.1 | - | 12 | 1927 | 62 | ||
| AK394797.1 | - | 12 | 1913 | 62 | ||
More Information
Set Membership: |
|

GUSB
Genomic Map