Assay Details
| Gene Symbol: RAB34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123178.1 | NP_001116650.1 | 9 | 652 | 92 | |
| XM_013989962.2 | XP_013845416.1 | 995 | 92 | |||
| XM_013989963.2 | XP_013845417.1 | 1280 | 92 | |||
| XM_013989965.2 | XP_013845419.1 | 1072 | 92 | |||
| XM_013989966.2 | XP_013845420.1 | 990 | 92 | |||
| XM_013989967.2 | XP_013845421.1 | 1275 | 92 | |||
| GenBank mRNA | AK231635.1 | - | 10 | 948 | 92 | |
| AK236728.1 | - | 10 | 856 | 92 | ||
| AK347296.1 | - | 10 | 929 | 92 | ||
| AK389435.1 | - | 10 | 869 | 92 | ||
| AK395489.1 | - | 10 | 969 | 92 | ||
| AY610189.1 | - | 7 | 681 | 92 | ||
| DQ917631.1 | - | 9 | 652 | 92 | ||
More Information
Set Membership: |
|

RAB34
Genomic Map