Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214016.2 | NP_999181.1 | 8 - 9 | 1453 | 64 | |
| GenBank mRNA | AF384216.1 | - | 8 - 9 | 1466 | 64 | |
| AK350440.1 | - | 9 - 10 | 1507 | 64 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map