Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044584.1 | NP_001038049.1 | 8 - 9 | 850 | 140 | |
| XM_021093604.1 | XP_020949263.1 | 1246 | 140 | |||
| XM_021093605.1 | XP_020949264.1 | 716 | 140 | |||
| GenBank mRNA | AK392064.1 | - | 7 - 8 | 815 | 140 | |
| DQ499448.1 | - | 8 - 9 | 850 | 140 | ||
| DQ841573.1 | - | 6 - 7 | 739 | 140 | ||
More Information
Set Membership: |
|

CRYZ
Genomic Map