Assay Details
| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077213.2 | NP_001070681.1 | 2 - 3 | 328 | 58 | |
| GenBank mRNA | AB529860.1 | - | 2 - 3 | 282 | 58 | |
| AF176246.1 | - | 2 - 3 | 282 | 58 | ||
| AK233883.1 | - | 2 - 3 | 354 | 58 | ||
| AK238500.1 | - | 2 - 3 | 354 | 58 | ||
| AK345303.1 | - | 2 - 3 | 354 | 58 | ||
| AK346279.1 | - | 2 - 3 | 354 | 58 | ||
| AK351290.1 | - | 2 - 3 | 354 | 58 | ||
| AK392500.1 | - | 2 - 3 | 376 | 58 | ||
| DQ989235.2 | - | 2 - 3 | 328 | 58 | ||
More Information
Set Membership: |
|

MIF
Genomic Map