Assay Details
| Gene Symbol: SCGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077224.1 | NP_001070692.1 | 10 - 11 | 706 | 86 | |
| XM_005665639.3 | XP_005665696.1 | 1222 | 86 | |||
| GenBank mRNA | AK237859.1 | - | 10 - 11 | 853 | 86 | |
| DQ972967.1 | - | 10 - 11 | 706 | 86 | ||
More Information
Set Membership: |
|

SCGN
Genomic Map