Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078680.1 | NP_001072148.1 | 2 - 3 | 175 | 125 | |
| XM_005656039.3 | XP_005656096.1 | 341 | 125 | |||
| XM_005656040.3 | XP_005656097.1 | 342 | 125 | |||
| GenBank mRNA | AK231866.1 | - | 3 - 4 | 256 | 125 | |
| AK343699.1 | - | 2 - 3 | 238 | 125 | ||
| AK396252.1 | - | 3 - 4 | 305 | 125 | ||
| AK399563.1 | - | 3 - 4 | 326 | 125 | ||
| AY610462.1 | - | 2 - 3 | 229 | 125 | ||
| DQ915496.1 | - | 2 - 3 | 175 | 125 | ||
More Information
Set Membership: |
|

CDC42
Genomic Map