Assay Details
| Gene Symbol: LDB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078685.1 | NP_001072153.1 | 10 - 11 | 1014 | 89 | |
| GenBank mRNA | AB242619.1 | - | 10 - 11 | 1014 | 89 | |
More Information
Set Membership: |
|

LDB1
Genomic Map