Assay Details
| Gene Symbol: APOBEC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097446.1 | NP_001090915.1 | 5 - 6 | 862 | 109 | |
| GenBank mRNA | AK240267.1 | - | 5 - 6 | 1007 | 109 | |
| AK350289.1 | - | 5 - 6 | 849 | 109 | ||
| DQ974647.1 | - | 5 - 6 | 862 | 109 | ||
| EU864540.1 | - | 2 - 3 | 306 | 109 | ||
| EU871586.1 | - | 5 - 6 | 1257 | 109 | ||
| HM347451.1 | - | 2 - 3 | 321 | 109 | ||
More Information
Set Membership: |
|

APOBEC3B
Genomic Map