Assay Details
| Gene Symbol: RPS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097481.2 | NP_001090950.1 | 3 - 4 | 334 | 71 | |
| GenBank mRNA | AK237453.1 | - | 3 - 4 | 334 | 71 | |
| AK392786.1 | - | 3 - 4 | 335 | 71 | ||
| AK399720.1 | - | 3 - 4 | 336 | 71 | ||
| AY610367.1 | - | 2 - 3 | 315 | 71 | ||
| AY610469.1 | - | 2 - 3 | 317 | 71 | ||
| DQ629173.1 | - | 3 - 4 | 335 | 71 | ||
More Information
Set Membership: |
|

RPS26
Genomic Map