Assay Details
| Gene Symbol: COX7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097484.1 | NP_001090953.1 | 3 - 4 | 278 | 113 | |
| GenBank mRNA | AK391921.1 | - | 3 - 4 | 323 | 113 | |
| DQ629178.1 | - | 3 - 4 | 278 | 113 | ||
More Information
Set Membership: |
|

COX7A2
Genomic Map