Assay Details
| Gene Symbol: BTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097505.2 | NP_001090974.1 | 1 - 2 | 264 | 89 | |
| GenBank mRNA | EF486867.1 | - | 1 - 2 | 277 | 89 | |
| EU255256.1 | - | 1 - 2 | 264 | 89 | ||
More Information
Set Membership: |
|

BTG2
Genomic Map