Assay Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099934.1 | NP_001093404.1 | 6 - 7 | 745 | 86 | |
| GenBank mRNA | AK237945.1 | - | 6 - 7 | 787 | 86 | |
| AK237946.1 | - | 6 - 7 | 787 | 86 | ||
| EF369512.1 | - | 6 - 7 | 745 | 86 | ||
| EF503637.1 | - | 6 - 7 | 745 | 86 | ||
More Information
Set Membership: |
|

TNMD
Genomic Map