Assay Details
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105312.1 | NP_001098782.1 | 1 - 2 | 424 | 79 | |
GenBank mRNA | AK236796.1 | - | 1 - 2 | 425 | 79 | |
AK237708.1 | - | 1 - 2 | 425 | 79 | ||
AK349001.1 | - | 1 - 2 | 425 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
