Assay Details
| Gene Symbol: RBM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105312.1 | NP_001098782.1 | 2 - 3 | 550 | 102 | |
| GenBank mRNA | AK236796.1 | - | 2 - 3 | 551 | 102 | |
| AK237708.1 | - | 2 - 3 | 551 | 102 | ||
| AK349001.1 | - | 2 - 3 | 551 | 102 | ||
More Information
Set Membership: |
|

RBM38
Genomic Map