Assay Details
| Gene Symbol: CIDEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112689.1 | NP_001106159.2 | 5 - 6 | 599 | 61 | |
| XM_005669744.3 | XP_005669801.1 | 734 | 61 | |||
| XM_005669745.3 | XP_005669802.1 | 649 | 61 | |||
| XM_013981921.1 | XP_013837375.1 | 632 | 61 | |||
| XM_013981922.2 | XP_013837376.1 | 553 | 61 | |||
| GenBank mRNA | AK232875.1 | - | 5 - 6 | 611 | 61 | |
| AK390817.1 | - | 5 - 6 | 735 | 61 | ||
| EF374062.1 | - | 5 - 6 | 599 | 61 | ||
| EU395789.1 | - | 4 - 5 | 563 | 61 | ||
More Information
Set Membership: |
|

CIDEC
Genomic Map