Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113436.1 | NP_001106907.1 | 6 - 7 | 836 | 101 | |
| GenBank mRNA | AK237331.1 | - | 6 - 7 | 851 | 101 | |
| AK390078.1 | - | 6 - 7 | 898 | 101 | ||
| EU295558.1 | - | 6 - 7 | 836 | 101 | ||
More Information
Set Membership: |
|

LRPAP1
Genomic Map