Assay Details
| Gene Symbol: MUC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113440.1 | NP_001106911.1 | 2 - 3 | 1452 | 125 | |
| XM_005670133.3 | XP_005670190.1 | 1563 | 125 | |||
| GenBank mRNA | AK349055.1 | - | 2 - 3 | 1441 | 125 | |
| EU330422.1 | - | 2 - 3 | 1452 | 125 | ||
| EU330423.1 | - | 2 - 3 | 1452 | 125 | ||
More Information
Set Membership: |
|

MUC20
Genomic Map