Assay Details
| Gene Symbol: ECI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113443.1 | NP_001106914.1 | 9 - 10 | 1033 | 65 | |
| GenBank mRNA | AK232489.1 | - | 9 - 10 | 1032 | 65 | |
| AK346380.1 | - | 9 - 10 | 1000 | 65 | ||
| AK393523.1 | - | 8 - 9 | 897 | 65 | ||
| DQ291159.1 | - | 9 - 10 | 1033 | 65 | ||
More Information
Set Membership: |
|

ECI2
Genomic Map