Assay Details
| Gene Symbol: SLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113695.1 | NP_001107167.1 | 1 - 2 | 175 | 99 | |
| GenBank mRNA | AB215120.1 | - | 1 - 2 | 103 | 99 | |
| AF464046.2 | - | 1 - 2 | 96 | 99 | ||
| AK348716.1 | - | 1 - 2 | 160 | 99 | ||
| AK394000.1 | - | 1 - 2 | 168 | 99 | ||
| AK396034.1 | - | 1 - 2 | 167 | 99 | ||
| AY135579.1 | - | 1 - 2 | 175 | 99 | ||
| DQ272492.1 | - | 1 - 2 | 103 | 99 | ||
| DQ883227.1 | - | 1 - 2 | 124 | 99 | ||
| EU432072.1 | - | 1 - 2 | 105 | 99 | ||
| EU432076.1 | - | 1 - 2 | 105 | 99 | ||
| EU432079.1 | - | 1 - 2 | 105 | 99 | ||
More Information
Set Membership: |
|

SLA-DRB1
Genomic Map