Assay Details
| Gene Symbol: SLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113695.1 | NP_001107167.1 | 4 - 5 | 840 | 117 | |
| GenBank mRNA | AB016746.1 | - | 3 - 4 | 681 | 117 | |
| AF464046.2 | - | 4 - 5 | 761 | 117 | ||
| AK348716.1 | - | 4 - 5 | 825 | 117 | ||
| AK394000.1 | - | 4 - 5 | 833 | 117 | ||
| AK396034.1 | - | 4 - 5 | 832 | 117 | ||
| AK396070.1 | - | 4 - 5 | 836 | 117 | ||
| AY135576.1 | - | 3 - 4 | 478 | 117 | ||
| AY135579.1 | - | 4 - 5 | 840 | 117 | ||
| AY135580.1 | - | 4 - 5 | 728 | 117 | ||
| DQ883227.1 | - | 4 - 5 | 789 | 117 | ||
| EU432072.1 | - | 4 - 5 | 770 | 117 | ||
| EU432076.1 | - | 4 - 5 | 770 | 117 | ||
| EU432079.1 | - | 4 - 5 | 770 | 117 | ||
| KU754598.1 | - | 4 - 5 | 789 | 117 | ||
| LC020542.1 | - | 4 - 5 | 768 | 117 | ||
| LC020543.1 | - | 4 - 5 | 768 | 117 | ||
| Gene Symbol: LOC106504372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK396361.1 | - | 4 - 5 | 858 | 117 | |
More Information
Set Membership: |
|

SLA-DRB1
Genomic Map