Assay Details
| Gene Symbol: SLA-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113703.1 | NP_001107175.1 | 5 - 6 | 980 | 74 | |
| XM_013977617.2 | XP_013833071.1 | 1158 | 74 | |||
| GenBank mRNA | AK231632.1 | - | 5 - 6 | 1026 | 74 | |
| AK235295.1 | - | 5 - 6 | 998 | 74 | ||
| AK348807.1 | - | 5 - 6 | 1026 | 74 | ||
| AK348811.1 | - | 2 - 3 | 1275 | 74 | ||
| AK389292.1 | - | 5 - 6 | 1032 | 74 | ||
| AK397717.1 | - | 4 - 5 | 1278 | 74 | ||
| AY463539.1 | - | 2 - 3 | 229 | 74 | ||
| AY463542.1 | - | 5 - 6 | 980 | 74 | ||
| GU322909.1 | - | 5 - 6 | 980 | 74 | ||
More Information
Set Membership: |
|

SLA-8
Genomic Map