Assay Details
| Gene Symbol: SLA-DQA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114062.2 | NP_001107534.2 | 4 - 5 | 820 | 84 | |
| GenBank mRNA | AK238471.1 | - | 4 - 5 | 823 | 84 | |
| AK393954.1 | - | 4 - 5 | 839 | 84 | ||
| AK397673.1 | - | 4 - 5 | 858 | 84 | ||
| AK397819.1 | - | 4 - 5 | 838 | 84 | ||
| AY285931.1 | - | 4 - 5 | 820 | 84 | ||
| M29938.1 | - | 4 - 5 | 798 | 84 | ||
More Information
Set Membership: |
|

SLA-DQA1
Genomic Map