Assay Details
| Gene Symbol: SLA-DOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114064.2 | NP_001107536.2 | 1 - 2 | 147 | 103 | |
| XM_021097880.1 | XP_020953539.1 | 434 | 103 | |||
| GenBank mRNA | AK397732.1 | - | 1 - 2 | 150 | 103 | |
| DQ333334.1 | - | 1 - 2 | 147 | 103 | ||
More Information
Set Membership: |
|

SLA-DOB
Genomic Map