Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122990.1 | NP_001116462.1 | 2 - 3 | 175 | 65 | |
| GenBank mRNA | AK349258.1 | - | 2 - 3 | 249 | 65 | |
| AK349448.1 | - | 2 - 3 | 244 | 65 | ||
| AK392342.1 | - | 2 - 3 | 246 | 65 | ||
| AK397125.1 | - | 2 - 3 | 244 | 65 | ||
| AK399142.1 | - | 2 - 3 | 104 | 65 | ||
| AK399155.1 | - | 2 - 3 | 248 | 65 | ||
| AK399175.1 | - | 2 - 3 | 244 | 65 | ||
More Information
Set Membership: |
|

NNAT
Genomic Map