Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123078.1 | NP_001116550.1 | 2 - 3 | 495 | 86 | |
| XM_021090887.1 | XP_020946546.1 | 1211 | 86 | |||
| GenBank mRNA | AK233046.1 | - | 2 - 3 | 503 | 86 | |
| AK233477.1 | - | 2 - 3 | 501 | 86 | ||
| AK238726.1 | - | 2 - 3 | 502 | 86 | ||
| AK392772.1 | - | 2 - 3 | 506 | 86 | ||
| AK394164.1 | - | 2 - 3 | 502 | 86 | ||
| AK398876.1 | - | 2 - 3 | 505 | 86 | ||
| EF062580.2 | - | 2 - 3 | 495 | 86 | ||
| HM991499.1 | - | 1 - 2 | 220 | 86 | ||
More Information
Set Membership: |
|

ATF4
Genomic Map