Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123143.1 | NP_001116615.1 | 5 - 6 | 707 | 120 | |
| XM_005664831.3 | XP_005664888.1 | 1390 | 120 | |||
| GenBank mRNA | AB516285.1 | - | 5 - 6 | 737 | 120 | |
| AK396179.1 | - | 5 - 6 | 736 | 120 | ||
More Information
Set Membership: |
|

NCR1
Genomic Map