Assay Details
| Gene Symbol: NDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123144.1 | NP_001116616.1 | 1 | 654 | 105 | |
| GenBank mRNA | AK351087.1 | - | 1 | 740 | 105 | |
| AK396929.1 | - | 1 | 741 | 105 | ||
| DQ917623.1 | - | 1 | 654 | 105 | ||
More Information
Set Membership: |
|

NDN
Genomic Map