Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123176.1 | NP_001116648.1 | 2 - 3 | 532 | 69 | |
| GenBank mRNA | AK238476.1 | - | 2 - 3 | 755 | 69 | |
| AK394570.1 | - | 2 - 3 | 755 | 69 | ||
| DQ917630.1 | - | 2 - 3 | 532 | 69 | ||
More Information
Set Membership: |
|

RAB32
Genomic Map