Assay Details
| Gene Symbol: AURKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213919.2 | NP_999084.2 | 5 - 6 | 513 | 93 | |
| XM_013990048.2 | XP_013845502.2 | 2275 | 93 | |||
| XM_013990050.2 | XP_013845504.2 | 1238 | 93 | |||
| XM_021066188.1 | XP_020921847.1 | 1310 | 93 | |||
| XM_021066189.1 | XP_020921848.1 | 1302 | 93 | |||
| XM_021066190.1 | XP_020921849.1 | 1077 | 93 | |||
| XM_021066191.1 | XP_020921850.1 | 1230 | 93 | |||
| XM_021066192.1 | XP_020921851.1 | 1038 | 93 | |||
| XM_021066193.1 | XP_020921852.1 | 1021 | 93 | |||
| XM_021066195.1 | XP_020921854.1 | 1069 | 93 | |||
| XM_021066196.1 | XP_020921855.1 | 2186 | 93 | |||
| XM_021066197.1 | XP_020921856.1 | 2191 | 93 | |||
| XM_021066198.1 | XP_020921857.1 | 2193 | 93 | |||
| GenBank mRNA | AK234554.1 | - | 5 - 6 | 513 | 93 | |
| AK236175.1 | - | 5 - 6 | 498 | 93 | ||
| AK238338.1 | - | 6 - 7 | 543 | 93 | ||
| AK398886.1 | - | 5 - 6 | 470 | 93 | ||
| AY336974.1 | - | 4 - 5 | 420 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021066197.1 transcripts.
Set Membership: |
|

AURKB
Genomic Map