Assay Details
| Gene Symbol: IFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213948.1 | NP_999113.1 | 3 - 4 | 390 | 68 | |
| GenBank mRNA | AF493993.1 | - | 3 - 4 | 375 | 68 | |
| AY188090.1 | - | 3 - 4 | 390 | 68 | ||
| AY293733.1 | - | 3 - 4 | 375 | 68 | ||
| AY562551.2 | - | 1 - 2 | 49 | 68 | ||
| DQ666352.1 | - | 3 - 4 | 366 | 68 | ||
| DQ839398.1 | - | 3 - 4 | 390 | 68 | ||
| DQ902588.1 | - | 3 - 4 | 68 | |||
| DQ913893.2 | - | 3 - 4 | 433 | 68 | ||
| EU249804.1 | - | 3 - 4 | 366 | 68 | ||
| GU433229.1 | - | 3 - 4 | 366 | 68 | ||
| JF906510.1 | - | 3 - 4 | 366 | 68 | ||
More Information
Set Membership: |
|

IFNG
Genomic Map