Assay Details
| Gene Symbol: HADHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348972.1 | NP_001335901.1 | 10 - 11 | 1071 | 63 | |
| GenBank mRNA | AF028007.1 | - | 7 - 8 | 835 | 63 | |
| AK239646.1 | - | 10 - 11 | 1018 | 63 | ||
| AK343679.1 | - | 10 - 11 | 1031 | 63 | ||
| AK389670.1 | - | 10 - 11 | 1039 | 63 | ||
| AK393956.1 | - | 10 - 11 | 1007 | 63 | ||
| AK400872.1 | - | 10 - 11 | 992 | 63 | ||
More Information
Set Membership: |
|

HADHB
Genomic Map