Assay Details
| Gene Symbol: IL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214029.1 | NP_999194.1 | 3 - 4 | 322 | 70 | |
| XM_005655198.3 | XP_005655255.1 | 463 | 70 | |||
| XM_013987856.2 | XP_013843310.1 | 642 | 70 | |||
| XM_013987857.2 | XP_013843311.1 | 540 | 70 | |||
| XM_021085846.1 | XP_020941505.1 | 1543 | 70 | |||
| GenBank mRNA | M86730.2 | - | 4 - 5 | 358 | 70 | |
| X52731.1 | - | 356 | 70 | |||
More Information
Set Membership: |
|

IL1A
Genomic Map