Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001644.1 | NP_001001644.1 | 1 - 2 | 267 | 64 | |
| XM_013986927.2 | XP_013842381.1 | 242 | 64 | |||
| GenBank mRNA | AK234238.1 | - | 1 - 2 | 266 | 64 | |
| AK349706.1 | - | 1 - 2 | 217 | 64 | ||
| AY553926.1 | - | 1 - 2 | 267 | 64 | ||
| AY609761.1 | - | 1 - 2 | 205 | 64 | ||
More Information
Set Membership: |
|

MED27
Genomic Map