Assay Details
| Gene Symbol: SLC52A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004033.1 | NP_001004033.1 | 1 - 2 | 127 | 67 | |
| XM_005662796.3 | XP_005662853.1 | 724 | 67 | |||
| XM_013996382.2 | XP_013851836.1 | 468 | 67 | |||
| XM_021088739.1 | XP_020944398.1 | 403 | 67 | |||
| XM_021088740.1 | XP_020944399.1 | 321 | 67 | |||
| GenBank mRNA | AK238538.1 | - | 2 - 3 | 590 | 67 | |
| AK351148.1 | - | 2 - 3 | 590 | 67 | ||
| AY134475.1 | - | 1 - 2 | 127 | 67 | ||
More Information
Set Membership: |
|

SLC52A2
Genomic Map