Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005350.2 | NP_001005350.1 | 10 - 11 | 1357 | 66 | |
| XM_013981457.2 | XP_013836911.1 | 1526 | 66 | |||
| XM_021068594.1 | XP_020924253.1 | 1504 | 66 | |||
| GenBank mRNA | AY705387.1 | - | 10 - 11 | 1357 | 66 | |
More Information
Set Membership: |
|

ACVR2B
Genomic Map