Assay Details
| Gene Symbol: MBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007194.3 | NP_001007195.1 | 3 - 4 | 345 | 120 | |
| GenBank mRNA | AY771222.2 | - | 3 - 4 | 345 | 120 | |
| EU421730.1 | - | 3 - 4 | 345 | 120 | ||
| HQ329180.1 | - | 2 - 3 | 304 | 120 | ||
| HQ329181.1 | - | 2 - 3 | 304 | 120 | ||
| HQ329182.1 | - | 2 - 3 | 304 | 120 | ||
More Information
Set Membership: |
|

MBL1
Genomic Map