Assay Details
| Gene Symbol: CA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008688.1 | NP_001008688.1 | 6 - 7 | 724 | 61 | |
| GenBank mRNA | AK233187.1 | - | 6 - 7 | 735 | 61 | |
| AK233384.1 | - | 6 - 7 | 778 | 61 | ||
| AK346298.1 | - | 5 - 6 | 650 | 61 | ||
| AK394144.1 | - | 6 - 7 | 717 | 61 | ||
| AK394364.1 | - | 6 - 7 | 870 | 61 | ||
| AK394623.1 | - | 6 - 7 | 630 | 61 | ||
| AK394692.1 | - | 6 - 7 | 727 | 61 | ||
| AK399490.1 | - | 6 - 7 | 712 | 61 | ||
| AK399512.1 | - | 6 - 7 | 711 | 61 | ||
| AK399626.1 | - | 6 - 7 | 712 | 61 | ||
| AY789514.1 | - | 6 - 7 | 685 | 61 | ||
| AY789645.1 | - | 6 - 7 | 724 | 61 | ||
More Information
Set Membership: |
|

CA3
Genomic Map