Assay Details
| Gene Symbol: GUCY1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018032.2 | NP_001018042.1 | 11 - 12 | 1712 | 102 | |
| XM_021100491.1 | XP_020956150.1 | 1826 | 102 | |||
| XM_021100492.1 | XP_020956151.1 | 1858 | 102 | |||
| XM_021100493.1 | XP_020956152.1 | 1944 | 102 | |||
| XM_021100494.1 | XP_020956153.1 | 1613 | 102 | |||
| XM_021100495.1 | XP_020956154.1 | 1639 | 102 | |||
| GenBank mRNA | AK390507.1 | - | 12 - 13 | 1805 | 102 | |
| AY661709.1 | - | 3 - 4 | 458 | 102 | ||
| DQ008578.1 | - | 11 - 12 | 1714 | 102 | ||
More Information
Set Membership: |
|

GUCY1B1
Genomic Map