Assay Details
| Gene Symbol: H1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025216.1 | NP_001020387.1 | 1 | 46 | 69 | |
| GenBank mRNA | AY489287.1 | - | 1 | 46 | 69 | |
| Gene Symbol: ABRACL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK239317.1 | - | 87 | 69 | ||
More Information
Set Membership: |
|

H1-2
Genomic Map