Assay Details
| Gene Symbol: APOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040640.1 | NP_001035730.1 | 1 - 2 | 169 | 75 | |
| XM_021098346.1 | XP_020954005.1 | 2040 | 75 | |||
| GenBank mRNA | AY610077.1 | - | 1 - 2 | 202 | 75 | |
| DQ329240.2 | - | 1 - 2 | 169 | 75 | ||
More Information
Set Membership: |
|

APOM
Genomic Map